Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007118.3 | NP_009049.2 | 55 - 56 | 8641 | 122 |
NR_134469.1 | 55 - 56 | 8111 | 122 | ||
XM_011514107.2 | XP_011512409.1 | 55 - 56 | 9216 | 122 | |
XM_011514108.1 | XP_011512410.1 | 55 - 56 | 8515 | 122 | |
XM_011514109.2 | XP_011512411.1 | 55 - 56 | 12293 | 122 | |
XM_011514110.2 | XP_011512412.1 | 55 - 56 | 8751 | 122 | |
XM_017009801.1 | XP_016865290.1 | 55 - 56 | 8981 | 122 | |
XM_017009803.1 | XP_016865292.1 | 48 - 49 | 7262 | 122 | |
GenBank mRNA | AF091395.1 | - | 55 - 56 | 8506 | 122 |
AK131423.1 | - | 8 - 9 | 1607 | 122 | |
BC017268.2 | - | 4 - 5 | 477 | 122 | |
U42390.1 | - | 55 - 56 | 7975 | 122 |