Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007118.3 | NP_009049.2 | 56 - 57 | 8775 | 82 |
NR_134469.1 | 56 - 57 | 8245 | 82 | ||
XM_011514107.2 | XP_011512409.1 | 56 - 57 | 9350 | 82 | |
XM_011514108.1 | XP_011512410.1 | 56 - 57 | 8649 | 82 | |
XM_011514109.2 | XP_011512411.1 | 56 - 57 | 12427 | 82 | |
XM_011514110.2 | XP_011512412.1 | 56 - 57 | 8885 | 82 | |
XM_017009801.1 | XP_016865290.1 | 56 - 57 | 9115 | 82 | |
XM_017009803.1 | XP_016865292.1 | 49 - 50 | 7396 | 82 | |
GenBank mRNA | AF091395.1 | - | 56 - 57 | 8640 | 82 |
AK131423.1 | - | 9 - 10 | 1741 | 82 | |
BC017268.2 | - | 5 - 6 | 611 | 82 | |
U42390.1 | - | 56 - 57 | 8109 | 82 |