Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032520.4 | NP_115909.1 | 8 - 9 | 79 | |
XM_017023782.1 | XP_016879271.1 | 8 - 9 | 79 | ||
XM_017023783.1 | XP_016879272.1 | 6 - 7 | 79 | ||
GenBank mRNA | AF302786.1 | - | 8 - 9 | 79 | |
AK312067.1 | - | 8 - 9 | 79 | ||
AY203933.1 | - | 8 - 9 | 79 | ||
BC014592.1 | - | 8 - 9 | 79 | ||
BC131525.1 | - | 7 - 8 | 79 | ||
BX398427.1 | - | 3 - 4 | 555 | 79 |