Assay Design



Gene Transcripts

Gene Symbol
KIAA0141
Entrez Gene ID
9812
Gene Name
KIAA0141
Gene Aliases
DELE
Chromosome Location
Chr.5: 141923785 - 141942047 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142603.2 NP_001136075.1 1 - 2 156 81
NM_014773.4 NP_055588.3 1 - 2 156 81
XM_005268547.3 XP_005268604.1 1 - 2 191 81
XM_005268548.3 XP_005268605.1 1 - 2 191 81
XM_005268552.2 XP_005268609.1 1 - 2 191 81
XM_011537715.1 XP_011536017.1 1 - 2 191 81
XM_011537716.2 XP_011536018.1 1 - 2 191 81
GenBank mRNA AK292869.1 - 1 - 2 105 81
BC007855.2 - 1 - 2 132 81
BC010452.1 - 1 - 2 47 81
BC011269.1 - 1 - 2 47 81
BC011926.1 - 1 - 2 77 81
BC035486.1 - 1 - 2 72 81
CN307416.1 - 1 - 2 192 81
D50931.1 - 1 - 2 114 81

View Full Product Details