Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170753.1 | NP_001164224.1 | 1 - 2 | 104 | 79 |
NM_145017.2 | NP_659454.2 | 1 - 2 | 104 | 79 | |
GenBank mRNA | AK057333.1 | - | 1 - 2 | 97 | 79 |
BC043246.1 | - | 1 - 2 | 102 | 79 | |
BI561157.1 | - | 1 - 2 | 102 | 79 | |
JF442033.1 | - | 1 - 2 | 77 | 79 |