Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017519.2 | NP_059989.2 | 10 - 11 | 3103 | 62 |
NM_020732.3 | NP_065783.3 | 11 - 12 | 3142 | 62 | |
XM_017011105.1 | XP_016866594.1 | 13 - 14 | 3389 | 62 | |
XM_017011106.1 | XP_016866595.1 | 12 - 13 | 3259 | 62 | |
GenBank mRNA | AF253515.1 | - | 10 - 11 | 2297 | 62 |
AF259792.1 | - | 6 - 7 | 1045 | 62 | |
AF468300.1 | - | 11 - 12 | 1672 | 62 | |
AF521671.1 | - | 10 - 11 | 2890 | 62 |