Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017519.2 | NP_059989.2 | 1 - 2 | 1542 | 86 |
NM_020732.3 | NP_065783.3 | 1 - 2 | 1542 | 86 | |
XM_005267069.4 | XP_005267126.2 | 1 - 2 | 1617 | 86 | |
XM_011535984.2 | XP_011534286.2 | 1 - 2 | 1618 | 86 | |
XM_017011103.1 | XP_016866592.1 | 1 - 2 | 1618 | 86 | |
XM_017011104.1 | XP_016866593.1 | 1 - 2 | 1617 | 86 | |
XM_017011105.1 | XP_016866594.1 | 1 - 2 | 1618 | 86 | |
XM_017011106.1 | XP_016866595.1 | 1 - 2 | 1617 | 86 | |
XM_017011107.1 | XP_016866596.1 | 1 - 2 | 1618 | 86 | |
XM_017011108.1 | XP_016866597.1 | 1 - 2 | 1625 | 86 | |
GenBank mRNA | AF253515.1 | - | 1 - 2 | 736 | 86 |
AF468300.1 | - | 1 - 2 | 72 | 86 | |
AF521671.1 | - | 1 - 2 | 1329 | 86 |