Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017519.2 | NP_059989.2 | 5 - 6 | 2237 | 59 |
NM_020732.3 | NP_065783.3 | 6 - 7 | 2276 | 59 | |
XM_005267069.4 | XP_005267126.2 | 5 - 6 | 2312 | 59 | |
XM_011535984.2 | XP_011534286.2 | 7 - 8 | 2394 | 59 | |
XM_017011103.1 | XP_016866592.1 | 7 - 8 | 2394 | 59 | |
XM_017011104.1 | XP_016866593.1 | 7 - 8 | 2393 | 59 | |
XM_017011105.1 | XP_016866594.1 | 7 - 8 | 2394 | 59 | |
XM_017011106.1 | XP_016866595.1 | 7 - 8 | 2393 | 59 | |
XM_017011107.1 | XP_016866596.1 | 7 - 8 | 2394 | 59 | |
XM_017011108.1 | XP_016866597.1 | 7 - 8 | 2401 | 59 | |
XM_017011109.1 | XP_016866598.1 | 3 - 4 | 4750 | 59 | |
GenBank mRNA | AF253515.1 | - | 5 - 6 | 1431 | 59 |
AF259792.1 | - | 1 - 2 | 179 | 59 | |
AF468300.1 | - | 6 - 7 | 806 | 59 | |
AF521671.1 | - | 5 - 6 | 2024 | 59 | |
AK025945.1 | - | 3 - 4 | 503 | 59 | |
AK074180.1 | - | 3 - 4 | 387 | 59 |