Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321115.1 | NP_001308044.1 | 1 - 2 | 545 | 128 |
NM_145023.5 | NP_659460.3 | 1 - 2 | 545 | 128 | |
NR_109827.1 | 1 - 2 | 545 | 128 | ||
GenBank mRNA | BC022020.2 | - | 1 - 2 | 451 | 128 |
BX647771.1 | - | 1 - 2 | 241 | 128 |