Assay Design



Gene Transcripts

Gene Symbol
ANKS1B
Entrez Gene ID
56899
Gene Name
ankyrin repeat and sterile alpha motif domain containing 1B
Gene Aliases
AIDA, AIDA-1, ANKS2, EB-1, EB1, cajalin-2
Chromosome Location
Chr.12: 98729904 - 99984727 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152788.4 NP_690001.3 3 - 4 782 103
XM_005269028.4 XP_005269085.1 3 - 4 393 103
XM_005269029.4 XP_005269086.1 3 - 4 365 103
XM_006719504.3 XP_006719567.1 3 - 4 855 103
XM_006719505.3 XP_006719568.1 3 - 4 855 103
XM_006719506.3 XP_006719569.1 3 - 4 855 103
XM_006719507.3 XP_006719570.1 3 - 4 855 103
XM_006719508.3 XP_006719571.1 3 - 4 855 103
XM_006719509.3 XP_006719572.1 3 - 4 855 103
XM_006719510.3 XP_006719573.1 3 - 4 855 103
XM_006719512.3 XP_006719575.1 3 - 4 855 103
XM_006719513.3 XP_006719576.1 3 - 4 855 103
XM_006719514.3 XP_006719577.1 3 - 4 855 103
XM_011538571.2 XP_011536873.1 3 - 4 855 103
XM_017019651.1 XP_016875140.1 3 - 4 855 103
XM_017019652.1 XP_016875141.1 3 - 4 855 103
XM_017019653.1 XP_016875142.1 3 - 4 855 103
XM_017019654.1 XP_016875143.1 3 - 4 855 103
XM_017019655.1 XP_016875144.1 3 - 4 855 103
XM_017019656.1 XP_016875145.1 3 - 4 855 103
XM_017019657.1 XP_016875146.1 3 - 4 855 103
XM_017019658.1 XP_016875147.1 3 - 4 855 103
GenBank mRNA AK124453.1 - 3 - 4 528 103
AY281131.1 - 3 - 4 365 103

View Full Product Details