Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006581.3 | NP_006572.2 | 3 - 3 | 2141 | 151 |
XM_011535383.2 | XP_011533685.1 | 3 - 3 | 7878 | 151 | |
XM_011535385.2 | XP_011533687.1 | 4 - 4 | 7995 | 151 | |
XM_017010188.1 | XP_016865677.1 | 4 - 4 | 5142 | 151 | |
XM_017010190.1 | XP_016865679.1 | 4 - 4 | 2068 | 151 | |
GenBank mRNA | AB023021.1 | - | 3 - 3 | 2094 | 151 |
BC036101.1 | - | 3 - 3 | 2118 | 151 |