Assay Design



Gene Transcripts

Gene Symbol
C16orf74
Entrez Gene ID
404550
Gene Name
chromosome 16 open reading frame 74
Gene Aliases
-
Chromosome Location
Chr.16: 85707517 - 85751083 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_206967.2 NP_996850.1 1 - 2 156 74
GenBank mRNA AB115764.1 - 1 - 2 142 74
CD674248.1 - 1 - 2 156 74

View Full Product Details