Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297619.1 | NP_001284548.1 | 4 - 5 | 359 | 148 |
NM_020917.2 | NP_065968.1 | 4 - 5 | 356 | 148 | |
XM_017027038.1 | XP_016882527.1 | 4 - 5 | 636 | 148 | |
GenBank mRNA | AB046779.1 | - | 3 - 4 | 329 | 148 |
AK293103.1 | - | 4 - 5 | 352 | 148 | |
AL699477.1 | - | 4 - 5 | 357 | 148 | |
BC144691.1 | - | 3 - 4 | 329 | 148 | |
BC152446.1 | - | 3 - 4 | 329 | 148 |