Assay Design



Gene Transcripts

Gene Symbol
YPEL2
Entrez Gene ID
388403
Gene Name
yippee like 2
Gene Aliases
FKSG4
Chromosome Location
Chr.17: 59331692 - 59401734 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005404.3 NP_001005404.1 1 - 2 129 150
GenBank mRNA BC132884.1 - 1 - 2 119 150
BC132886.1 - 1 - 2 119 150

View Full Product Details