Assay Design



Gene Transcripts

Gene Symbol
HES3
Entrez Gene ID
390992
Gene Name
hes family bHLH transcription factor 3
Gene Aliases
bHLHb43
Chromosome Location
Chr.1: 6244192 - 6245578 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024598.3 NP_001019769.1 3 - 4 228 88
GenBank mRNA CN370501.1 - 3 - 4 244 88
CX755241.1 - 3 - 4 226 88

View Full Product Details