Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300850.1 | NP_001287779.1 | 13 - 14 | 2611 | 58 |
NM_001300851.1 | NP_001287780.1 | 13 - 14 | 2611 | 58 | |
NM_001300852.1 | NP_001287781.1 | 13 - 14 | 2611 | 58 | |
NM_172071.3 | NP_742068.1 | 13 - 14 | 2611 | 58 | |
XM_005244921.3 | XP_005244978.1 | 13 - 14 | 2548 | 58 | |
XM_011509231.2 | XP_011507533.1 | 13 - 14 | 2611 | 58 | |
GenBank mRNA | AB095945.1 | - | 13 - 14 | 2452 | 58 |
AK093501.1 | - | 4 - 5 | 760 | 58 | |
AK122948.1 | - | 13 - 14 | 2611 | 58 | |
BC136784.1 | - | 13 - 14 | 2565 | 58 | |
BC144408.1 | - | 13 - 14 | 2565 | 58 |