Assay Design



Gene Transcripts

Gene Symbol
AIDA
Entrez Gene ID
64853
Gene Name
axin interactor, dorsalization associated
Gene Aliases
C1orf80
Chromosome Location
Chr.1: 222668013 - 222712522 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022831.2 NP_073742.2 1 - 2 304 84
GenBank mRNA AK022868.1 - 1 - 2 256 84
AK289865.1 - 1 - 2 255 84
AL833718.1 - 1 - 2 94 84
BC015535.1 - 1 - 2 231 84
BC067805.1 - 1 - 2 252 84
BC171848.1 - 1 - 2 279 84
DA483113.1 - 1 - 1 253 84

View Full Product Details