Assay Design



Gene Transcripts

Gene Symbol
HNRNPCL1
Entrez Gene ID
343069
Gene Name
heterogeneous nuclear ribonucleoprotein C-like 1
Gene Aliases
HNRPCL1
Chromosome Location
Chr.1: 12847377 - 12848756 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013631.2 NP_001013653.1 1 - 2 78 85
GenBank mRNA BC002696.2 - 1 - 2 47 85
BC137258.1 - 1 - 2 78 85
BX283835.1 - 1 - 2 48 85

View Full Product Details