Assay Design



Gene Transcripts

Gene Symbol
GARNL3
Entrez Gene ID
84253
Gene Name
GTPase activating Rap/RanGAP domain like 3
Gene Aliases
bA356B19.1
Chromosome Location
Chr.9: 127224471 - 127393655 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005252268.2 XP_005252325.1 1 - 2 221 71
XM_011519090.2 XP_011517392.1 1 - 2 66 71

View Full Product Details