Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_024346.3 | 10 - 11 | 79 | ||
GenBank mRNA | DY655576.1 | - | 1 - 2 | 55 | 79 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | maltase-glucoamylase 2 (putative) | MGAM2 | -17.2kb | Anti-sense | Chr. - 142111703 - 142223733 on Build GRCh38 | |
Downstream | protease, serine 58 | PRSS58 | PRSS1, TRY1, TRYX3, UNQ2540 | 9.8kb | Sense | Chr. - 142252138 - 142258058 on Build GRCh38 |