Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012267.2 | NP_001012267.1 | 2 - 3 | 810 | 97 |
XM_011518685.2 | XP_011516987.1 | 2 - 3 | 821 | 97 | |
XM_011518689.1 | XP_011516991.1 | 2 - 3 | 821 | 97 | |
XM_017014715.1 | XP_016870204.1 | 2 - 3 | 821 | 97 | |
GenBank mRNA | AK092603.1 | - | 2 - 3 | 625 | 97 |
BC071726.1 | - | 2 - 3 | 336 | 97 |