Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099289.2 | NP_001092759.1 | 9 - 10 | 2481 | 76 |
XM_011511110.2 | XP_011509412.1 | 7 - 8 | 2283 | 76 | |
GenBank mRNA | AK074131.1 | - | 7 - 8 | 1556 | 76 |
AL831825.1 | - | 2 - 3 | 469 | 76 |