Assay Design



Gene Transcripts

Gene Symbol
CHRNB4
Entrez Gene ID
1143
Gene Name
cholinergic receptor nicotinic beta 4 subunit
Gene Aliases
-
Chromosome Location
Chr.15: 78623282 - 78655586 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011521186.2 XP_011519488.1 3 - 4 201 62
XM_011521187.2 XP_011519489.1 2 - 3 108 62
GenBank mRNA AK310443.1 - 7 - 8 921 62

View Full Product Details