Assay Design



Gene Transcripts

Gene Symbol
SH2D7
Entrez Gene ID
646892
Gene Name
SH2 domain containing 7
Gene Aliases
-
Chromosome Location
Chr.15: 78089895 - 78104051 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101404.1 NP_001094874.1 5 - 6 1310 85

View Full Product Details