Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_002947.2 | 2 - 3 | 730 | 73 | |
GenBank mRNA | AB026156.1 | - | 2 - 3 | 730 | 73 |
LC005096.1 | - | 4 - 5 | 821 | 73 | |
LC005097.1 | - | 4 - 5 | 590 | 73 | |
LC005098.1 | - | 3 - 4 | 558 | 73 | |
LC005099.1 | - | 4 - 5 | 675 | 73 | |
LC005100.1 | - | 4 - 5 | 606 | 73 | |
LC005101.1 | - | 4 - 5 | 572 | 73 |