Assay Design



Gene Transcripts

Gene Symbol
KRT80
Entrez Gene ID
144501
Gene Name
keratin 80
Gene Aliases
KB20
Chromosome Location
Chr.12: 52168996 - 52192000 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081492.1 NP_001074961.1 3 - 4 672 76
NM_182507.2 NP_872313.2 3 - 4 672 76
GenBank mRNA AJ717743.1 - 3 - 4 626 76
BC065180.1 - 3 - 4 672 76

View Full Product Details