Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017971.2 | NP_001017971.1 | 7 - 8 | 1403 | 61 |
XM_011543723.2 | XP_011542025.1 | 7 - 8 | 2050 | 61 | |
XM_017010039.1 | XP_016865528.1 | 2 - 3 | 209 | 61 | |
XM_017010040.1 | XP_016865529.1 | 2 - 3 | 114 | 61 | |
XM_017010041.1 | XP_016865530.1 | 8 - 9 | 2128 | 61 | |
XM_017010042.1 | XP_016865531.1 | 9 - 10 | 2034 | 61 | |
XM_017010043.1 | XP_016865532.1 | 8 - 9 | 2117 | 61 | |
XM_017010044.1 | XP_016865533.1 | 7 - 8 | 2076 | 61 | |
XM_017010045.1 | XP_016865534.1 | 7 - 8 | 1628 | 61 | |
XM_017010046.1 | XP_016865535.1 | 7 - 8 | 1709 | 61 | |
XM_017010047.1 | XP_016865536.1 | 6 - 7 | 1998 | 61 | |
GenBank mRNA | AK022625.1 | - | 7 - 8 | 1403 | 61 |
BC093982.1 | - | 1 - 2 | 133 | 61 | |
BC112191.1 | - | 1 - 2 | 133 | 61 |