Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322255.1 | NP_001309184.1 | 2 - 3 | 667 | 70 |
GenBank mRNA | AK056573.1 | - | 1 - 2 | 84 | 70 |
BC101295.1 | - | 1 - 2 | 78 | 70 | |
BC101296.1 | - | 1 - 2 | 78 | 70 | |
BC101297.2 | - | 1 - 2 | 78 | 70 | |
BC101298.2 | - | 1 - 2 | 78 | 70 |