Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_027791.3 | 1 - 2 | 260 | 113 | |
NR_136543.1 | 1 - 2 | 260 | 113 | ||
GenBank mRNA | AL700268.1 | - | 1 - 2 | 171 | 113 |
CF135136.1 | - | 1 - 2 | 267 | 113 | |
CF135139.1 | - | 1 - 2 | 267 | 113 |