Assay Design
Gene Transcripts
- Gene Symbol
- MIR99AHG
- Entrez Gene ID
-
388815
- Gene Name
- mir-99a-let-7c cluster host gene
- Gene Aliases
-
C21orf34, C21orf35, LINC00478, MONC
- Chromosome Location
-
Chr.21: 16070488 - 16631727 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_136552.1 |
|
4 - 5 |
470 |
156 |
View Full Product Details