Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308025.1 | NP_001294954.1 | 3 - 4 | 1172 | 65 |
NM_152259.3 | NP_689472.3 | 3 - 4 | 1175 | 65 | |
GenBank mRNA | DR731358.1 | - | 4 - 5 | 65 | |
GQ227787.1 | - | 3 - 4 | 1172 | 65 |