Assay Design
Gene Transcripts
- Gene Symbol
- LOC283140
- Entrez Gene ID
-
283140
- Gene Name
- uncharacterized LOC283140
- Gene Aliases
-
-
- Chromosome Location
-
Chr.11: 112270749 - 112362534 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_126004.1 |
|
3 - 4 |
286 |
62 |
GenBank mRNA |
AK095275.1 |
- |
3 - 4 |
290 |
62 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DA440806.1 |
- |
3 - 4 |
290 |
|
Chr. 11 - 112270749 - 112360397 [+] on Build GRCh38 |
62 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
placenta expressed transcript 1 |
PLET1 |
C11orf34 |
-97.2kb
|
Anti-sense |
Chr. - 112248153 - 112260860 on Build GRCh38 |
Downstream |
uncharacterized LOC101928823 |
LOC101928823 |
|
119.7kb
|
Sense |
Chr. - 112482232 - 112541916 on Build GRCh38 |
View Full Product Details