Assay Design



Gene Transcripts

Gene Symbol
KIF6
Entrez Gene ID
221458
Gene Name
kinesin family member 6
Gene Aliases
C6orf102, dJ1043E3.1, dJ137F1.4, dJ188D3.1
Chromosome Location
Chr.6: 39335100 - 39725405 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289020.1 NP_001275949.1 2 - 3 264 88
NM_001289021.1 NP_001275950.1 2 - 3 264 88
NM_145027.4 NP_659464.3 2 - 3 264 88
XM_005248904.4 XP_005248961.1 2 - 3 264 88
XM_011514357.2 XP_011512659.1 2 - 3 189 88
XM_011514358.2 XP_011512660.1 2 - 3 189 88
XM_011514359.2 XP_011512661.1 2 - 3 189 88
XM_011514361.2 XP_011512663.1 2 - 3 264 88
XM_017010427.1 XP_016865916.1 2 - 3 264 88
GenBank mRNA AK131471.1 - 2 - 3 264 88
BC117307.1 - 2 - 3 194 88
BC143444.1 - 2 - 3 194 88
BC143445.1 - 2 - 3 194 88
DB080579.1 - 2 - 3 264 88

View Full Product Details