Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052846.1 | NP_443078.1 | 2 - 3 | 511 | 82 |
GenBank mRNA | AB089149.1 | - | 2 - 3 | 511 | 82 |
BC101043.1 | - | 2 - 3 | 287 | 82 | |
BC101044.2 | - | 2 - 3 | 287 | 82 | |
BC101045.1 | - | 2 - 3 | 287 | 82 | |
BC101046.1 | - | 2 - 3 | 287 | 82 |