Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032538.1 | NP_115927.1 | 8 - 9 | 1018 | 64 |
XM_011514947.2 | XP_011513249.1 | 7 - 8 | 808 | 64 | |
XM_017011364.1 | XP_016866853.1 | 7 - 8 | 808 | 64 | |
XM_017011365.1 | XP_016866854.1 | 7 - 8 | 808 | 64 | |
XM_017011366.1 | XP_016866855.1 | 7 - 8 | 808 | 64 | |
XM_017011367.1 | XP_016866856.1 | 7 - 8 | 808 | 64 | |
GenBank mRNA | AB058758.1 | - | 6 - 7 | 587 | 64 |
AB218664.1 | - | 9 - 10 | 822 | 64 | |
AK131217.1 | - | 7 - 8 | 748 | 64 |