Assay Design
Gene Transcripts
- Gene Symbol
- SH2D4B
- Entrez Gene ID
-
387694
- Gene Name
- SH2 domain containing 4B
- Gene Aliases
-
-
- Chromosome Location
-
Chr.10: 80537902 - 80646560 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK091518.1 |
- |
1 - 2 |
58 |
65 |
View Full Product Details