Assay Design
Gene Transcripts
- Gene Symbol
- ADAM6
- Entrez Gene ID
-
8755
- Gene Name
- ADAM metallopeptidase domain 6 (pseudogene)
- Gene Aliases
-
C14orf96, tMDCIV
- Chromosome Location
-
Chr.14: 105969731 - 105972269 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK093400.1 |
- |
1 - 2 |
579 |
90 |
View Full Product Details