Assay Design



Gene Transcripts

Gene Symbol
LRRC75A
Entrez Gene ID
388341
Gene Name
leucine rich repeat containing 75A
Gene Aliases
C17orf76, FAM211A
Chromosome Location
Chr.17: 16442005 - 16492191 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113567.2 NP_001107039.1 3 - 4 687 152
XM_017024619.1 XP_016880108.1 3 - 4 456 152
XM_017024620.1 XP_016880109.1 4 - 5 619 152
GenBank mRNA BC144381.1 - 3 - 4 406 152
BF966204.1 - 3 - 4 404 152

View Full Product Details