Assay Design



Gene Transcripts

Gene Symbol
HEPACAM2
Entrez Gene ID
253012
Gene Name
HEPACAM family member 2
Gene Aliases
MIKI
Chromosome Location
Chr.7: 93188586 - 93232297 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039372.2 NP_001034461.1 1 - 2 156 91
GenBank mRNA AK096002.1 - 1 - 2 109 91
DA426282.1 - 1 - 2 156 91

View Full Product Details