Assay Design
Gene Transcripts
- Gene Symbol
- HIGD2B
- Entrez Gene ID
-
123346
- Gene Name
- HIG1 hypoxia inducible domain family member 2B
- Gene Aliases
-
HIGD2BP
- Chromosome Location
-
Chr.15: 72675782 - 72686149 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002780.1 |
|
1 - 2 |
324 |
72 |
GenBank mRNA |
BC040890.1 |
- |
1 - 2 |
324 |
72 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BM564128.1 |
- |
1 - 2 |
344 |
|
Chr. 15 - 72675889 - 72686150 [-] on Build GRCh38 |
72 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
golgin A6 family member B |
GOLGA6B |
GOLGA, GOLGA6D |
-12.6kb
|
Anti-sense |
Chr. - 72638291 - 72669604 on Build GRCh38 |
Downstream |
Bardet-Biedl syndrome 4 |
BBS4 |
|
30.0
|
Anti-sense |
Chr. - 72686179 - 72738476 on Build GRCh38 |
View Full Product Details