Assay Design



Gene Transcripts

Gene Symbol
TCEAL5
Entrez Gene ID
340543
Gene Name
transcription elongation factor A like 5
Gene Aliases
WEX4
Chromosome Location
Chr.X: 103273690 - 103276869 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012979.2 NP_001012997.1 1 - 2 200 54
GenBank mRNA BC132922.1 - 1 - 2 77 54
BC132924.1 - 1 - 2 77 54
BM725918.1 - 1 - 2 200 54

View Full Product Details