Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199046.2 | NP_950247.2 | 4 - 5 | 664 | 111 |
NM_199456.2 | NP_955535.2 | 4 - 5 | 664 | 111 | |
GenBank mRNA | AY363222.1 | - | 4 - 5 | 664 | 111 |
AY363223.1 | - | 4 - 5 | 664 | 111 | |
BC104458.1 | - | 4 - 5 | 655 | 111 | |
BC104459.1 | - | 4 - 5 | 655 | 111 | |
BF980073.1 | - | 5 - 6 | 111 |