Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136106.4 | NP_001129578.1 | 2 - 3 | 404 | 64 |
NM_001178020.2 | NP_001171491.1 | 3 - 4 | 511 | 64 | |
NM_001197224.3 | NP_001184153.1 | 2 - 3 | 404 | 64 | |
NM_001197225.3 | NP_001184154.1 | 2 - 3 | 404 | 64 | |
XM_011522883.1 | XP_011521185.1 | 3 - 4 | 617 | 64 | |
XM_011522884.1 | XP_011521186.1 | 3 - 4 | 922 | 64 | |
XM_011522887.2 | XP_011521189.1 | 3 - 4 | 617 | 64 | |
XM_011522888.1 | XP_011521190.1 | 4 - 5 | 1118 | 64 | |
XM_011522889.1 | XP_011521191.1 | 3 - 4 | 933 | 64 | |
XM_011522890.1 | XP_011521192.1 | 4 - 5 | 1423 | 64 | |
XM_011522891.1 | XP_011521193.1 | 4 - 5 | 802 | 64 | |
XM_011522892.2 | XP_011521194.1 | 2 - 3 | 1071 | 64 | |
GenBank mRNA | AB472390.1 | - | 3 - 4 | 437 | 64 |
AB472391.1 | - | 2 - 3 | 330 | 64 | |
AB472392.1 | - | 2 - 3 | 330 | 64 | |
AB472394.1 | - | 1 - 2 | 157 | 64 | |
AB472396.1 | - | 2 - 3 | 330 | 64 | |
AK056142.1 | - | 2 - 3 | 364 | 64 | |
BC107477.1 | - | 1 - 2 | 266 | 64 |