Assay Design



Gene Transcripts

Gene Symbol
CENPW
Entrez Gene ID
387103
Gene Name
centromere protein W
Gene Aliases
C6orf173, CENP-W, CUG2
Chromosome Location
Chr.6: 126339696 - 126483320 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286525.1 NP_001273454.1 1 - 2 611 72
XM_017010845.1 XP_016866334.1 1 - 2 704 72
GenBank mRNA BM874602.1 - 1 - 2 212 72

View Full Product Details