Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181789.2 | NP_861454.2 | 1 - 2 | 528 | 62 |
XM_017022121.1 | XP_016877610.1 | 1 - 2 | 389 | 62 | |
XM_017022125.1 | XP_016877614.1 | 1 - 2 | 389 | 62 | |
GenBank mRNA | AK172756.1 | - | 1 - 2 | 364 | 62 |