Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_176818.2 | NP_789788.1 | 2 - 3 | 293 | 63 |
GenBank mRNA | AK094319.1 | - | 2 - 3 | 287 | 63 |
BC064523.1 | - | 2 - 3 | 264 | 63 | |
BC107145.1 | - | 2 - 3 | 250 | 63 | |
BC107146.1 | - | 2 - 3 | 243 | 63 |