Assay Design
Gene Transcripts
- Gene Symbol
- TTTY7
- Entrez Gene ID
-
246122
- Gene Name
- testis-specific transcript, Y-linked 7 (non-protein coding)
- Gene Aliases
-
CLONE795723, LINC00129, NCRNA00129, TTTY7A, TTTY7B, TTY7
- Chromosome Location
-
Chr.Y: 6449468 - 6457906 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_001534.2 |
|
5 - 6 |
863 |
139 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AF332233.3 |
- |
5 - 6 |
863 |
|
Chr. Y - 6449468 - 6457906 [+] on Build GRCh38 |
139 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
testis-specific transcript, Y-linked 21B (non-protein coding) |
TTTY21B |
|
-9.0kb
|
Sense |
Chr. - 6443434 - 6447077 on Build GRCh38 |
Downstream |
testis-specific transcript, Y-linked 8B (non-protein coding) |
TTTY8B |
NCRNA00217, TTTY8 |
13.6kb
|
Anti-sense |
Chr. - 6471031 - 6473630 on Build GRCh38 |
View Full Product Details