Assay Design



Gene Transcripts

Gene Symbol
HES5
Entrez Gene ID
388585
Gene Name
hes family bHLH transcription factor 5
Gene Aliases
bHLHb38
Chromosome Location
Chr.1: 2528745 - 2530554 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001010926.3 NP_001010926.1 2 - 3 303 107
GenBank mRNA DQ272660.1 - 2 - 3 303 107

View Full Product Details