Assay Design



Gene Transcripts

Gene Symbol
LGALS16
Entrez Gene ID
148003
Gene Name
galectin 16
Gene Aliases
-
Chromosome Location
Chr.19: 39655894 - 39660647 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190441.2 NP_001177370.2 3 - 4 371 82
GenBank mRNA BX095861.1 - 3 - 4 330 82
FJ613352.1 - 3 - 4 371 82

View Full Product Details