Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207352.3 | NP_997235.3 | 9 - 10 | 1522 | 75 |
XM_017008037.1 | XP_016863526.1 | 8 - 9 | 1280 | 75 | |
GenBank mRNA | AK022114.1 | - | 3 - 4 | 395 | 75 |
AK122600.1 | - | 9 - 10 | 1257 | 75 | |
AK126473.1 | - | 10 - 11 | 1191 | 75 | |
AK311390.1 | - | 3 - 4 | 308 | 75 | |
AY422002.1 | - | 9 - 10 | 1522 | 75 | |
BC041839.1 | - | 2 - 3 | 956 | 75 | |
BC060857.1 | - | 9 - 10 | 1388 | 75 | |
BX648537.1 | - | 3 - 4 | 453 | 75 | |
BX648730.1 | - | 4 - 5 | 5276 | 75 |